Tri-tert-butyl-1,4,7,10-tetraazacyclododecane-1,4,7,10-tetraacetate
Catalog #: Q-2525.0250BA
Supplier: Bachem Americas
CAS Number: 137076-54-1
Specifications
- Size:250 mg
- Cat. no.:Q-2525.0250BA
- Supplier No.:Q-2525.0250
Specifications
About this item
Synonyms: DOTA(OtBu)₃,2-(4,7,10-Tris(2-(tert-butoxy)-2-oxoethyl)-1,4,7,10-tetraazacyclododecan-1-yl)acetic acid,1,4,7,10-Tetraazacyclododecane-1,4,7,10-tetraacetic Acid Tri-tert-butyl Ester,4,7,10-Tri-(t-butyloxycarbonylmethyl)-1,4,7
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 572.74 g/mol
:Tri-tert-butyl-1,4,7,10-tetraazacyclododecane-1,4,7,10-tetraacetate
Formula:C₂₈H₅₂N₄O₈
MW:572.74 g/mol
Storage Temperature:Refrigerator
MDL Number:MFCD02259697
CAS Number:137076-54-1
Customers Who Bought This Also Bought