Benzyl N-(2-aminoethyl)carbamate hydrochloride
Supplier: Bachem Americas
Certificates
About this item
Synonyms: N-(2-Aminoethyl)carbamic acid benzyl ester hydrochloride,N-1-Z-1,2-diaminoethane hydrochloride,N-Z-Ethylenediamine hydrochloride,N-Carbobenzoxy-1,2-diaminoethane hydrochloride,N-Cbz-1
Formula:C₁₀H₁₄N₂O₂
MW:230.69 g/mol
MDL Number:MFCD00270150
CAS Number:72080-83-2
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 230.69 g/mol
Storage Temperature:Refrigerator
Specification Test Results
- Benzyl N-(2-aminoethyl)carbamate hydrochloride:
Specification Test Results
Customers Who Bought This Also Bought