mono-Fmoc ethylene diamine hydrochloride
Supplier: Bachem Americas
Certificates
About this item
Synonyms: (9H-Fluoren-9-yl)methyl (2-aminoethyl)carbamate hydrochloride,N-Fmoc-ethylenediamine hydrochloride
CAS Number:391624-46-7
:166410-32-8
Formula:C₁₇H₁₈N₂O₂·HCl
MW:318.80 g/mol
MDL Number:MFCD00830741
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 318.80 g/mol
Storage Temperature:Refrigerator
Specification Test Results
- mono-Fmoc ethylene diamine hydrochloride:
Specification Test Results
Customers Who Bought This Also Bought