D-Lactic acid benzyl ester
Supplier: Bachem Americas
Certificates
About this item
Formula:C₁₀H₁₂O₃
MDL Number:MFCD00672497
CAS Number:74094-05-6
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 180,20 g/mol
:D-Lactic acid benzyl ester
Storage Temperature:Refrigerator
Customers Who Bought This Also Bought