Xylazine
Catalog #: Q-1445.0025BA
Supplier: Bachem Americas
CAS Number: 7361-61-7
Specifications
- Size:25 g
- Cat. no.:Q-1445.0025BA
Specifications
About this item
Synonyms: Xylazine (free base)
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 220.334 g/mol
:Xylazine
Formula:C₁₂H₁₆N₂S
MW:220.334 g/mol
Storage Temperature:Freezer
MDL Number:MFCD00057908
CAS Number:7361-61-7
Customers Who Bought This Also Bought