Xylazine hydrochloride
Catalog #: Q-1440.0025BA
Supplier: Bachem Americas
CAS Number: 23076-35-9
Specifications
- Size:25 g
- Cat. no.:Q-1440.0025BA
- Supplier No.:Q-1440.0025
Specifications
About this item
Synonyms: N-(2,6-Dimethylphenyl)-5,6-dihydro-4H-1,3-thiazin-2-amine Hydrochloride,2-[(2,6-Dimethylphenyl)amino]-5,6-dihydro-4H-1
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 256.79 g/mol
:Xylazine hydrochloride
Formula:C₁₂H₁₆N₂S
MW:256.79 g/mol
Storage Temperature:Ambient
MDL Number:MFCD00058196
CAS Number:23076-35-9
Customers Who Bought This Also Bought