9-Fluorenylmethyl chloroformate (Fmoc chloride)
Catalog #: Q-1235.0025BA
Supplier: Bachem Americas
CAS Number: 28920-43-6
Specifications
- Size:25 g
- Cat. no.:Q-1235.0025BA
- Supplier No.:Q-1235.0025
Specifications
About this item
Synonyms: (9H-Fluoren-9-yl)methyl carbonochloridate,9-Fluorenylmethyl chloroformate,Fmoc-Cl,9H-Fluoren-9-ylmethyl chloroformate,Chloroformic acid 9-fluorenylmethyl ester
ADR:8,II
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 258.7 g/mol
UN:2923
:8
:II
Formula:C₁₅H₁₁ClO₂
MW:258.7 g/mol
Storage Temperature:Freezer
MDL Number:MFCD00001138
CAS Number:28920-43-6
Customers Who Bought This Also Bought