Dicarboxidine hydrochloride
Supplier: Bachem Americas
Certificates
About this item
Synonyms: Dicarboxidine·2 HCl
Formula:C₂₀H₂₆Cl₂N₂O₆
MW:461.336 g/mol
MDL Number:MFCD00236955
CAS Number:56455-90-4
:Dicarboxidine hydrochloride
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 461.336 g/mol
Storage Temperature:Refrigerator
Customers Who Bought This Also Bought