Ac-α-benzyl-4,6-O-benzylidene muramic acid
Supplier: Bachem Americas
Certificates
About this item
Synonyms: 2-(3-Acetamido-2-benzyl-4,6,O-benzylidene-alpha-D-glucopyranosid-4-yloxy)propanoic acid
Formula:C₂₅H₂₉NO₈
MW:471.506 g/mol
MDL Number:MFCD00057299
CAS Number:2862-03-5
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 471.506 g/mol
Storage Temperature:Refrigerator
Customers Who Bought This Also Bought