n-Dodecyl-β-D-maltoside
Supplier: Bachem Americas
Certificates
About this item
Synonyms: DDM,Lauryl-β-D-maltoside,Dodecyl-β-D-maltopyranoside,(2R,3R,4S,5S,6R)-2-(((2R,3S,4R,5R,6R)-6-(Dodecyloxy)-4,5-dihydroxy-2-(hydroxymethyl)tetrahydro-2H-pyran-3-yl)oxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol
Formula:C₂₄H₄₆O₁₁
MW:510.6 g/mol
MDL Number:MFCD00043012
CAS Number:69227-93-6
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 510.6 g/mol
Storage Temperature:Freezer
Customers Who Bought This Also Bought