1,2-Dilauroyl-sn-glycero-3-phosphocholine
Catalog #: O-1455.0001BA
Supplier: Bachem Americas
CAS Number: 18194-25-7
Specifications
- Size:1 g
- Cat. no.:O-1455.0001BA
- Supplier No.:O-1455.0001
Specifications
About this item
Synonyms: DLPC
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 621.837 g/mol
:1,2-Dilauroyl-sn-glycero-3-phosphocholine
Formula:C₃₂H₆₄NO₈P
MW:621.837 g/mol
Storage Temperature:Freezer
MDL Number:MFCD00066561
CAS Number:18194-25-7
Customers Who Bought This Also Bought