2-Aminoethyl-(R)-2,3-bis(hexadecyloxy)propyl hydrogen phosphate
Supplier: Bachem Americas
Certificates
About this item
Synonyms: 1,2-O-Dihexadecyl-sn-glycero-3-phosphoethanolamine
Formula:C₃₇H₇₈NO₆P
MW:664.006 g/mol
MDL Number:MFCD00042830
CAS Number:61423-61-8
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 664.006 g/mol
Storage Temperature:Freezer
Customers Who Bought This Also Bought