Fluorescein-6-carbonyl-Tyr-Val-Ala-DL-Asp(OMe)-fluoromethylketone
Catalog #: N-1960.1000BA
Supplier: Bachem Americas
CAS Number:
Specifications
- Size:1 mg
- Storage conditions:–20 ± 5 °C
- MDL No.:MFCD11975049
- Protein/peptide name:Fluorescein-6-carbonyl-Tyr-Val-Ala-DL-Asp(OMe)-fluoromethylketone
- Molecular weight:854.84
- Sequence:Fluorescein-6-carbonyl-Tyr-Val-Ala-DL-Asp(OMe)-fluoromethylketone
- Molecular formula:C₄₅H₅₀FN₅O₁₅
- Cat. no.:N-1960.1000BA
- Supplier No.:N-1960.1000
Specifications
About this item
See also the Asp derivatives N-1425 and N-1430, and Z-YVAD-FMK (N-1595).
6-FAM-YVAD(OMe)-FMK is a caspase-1 inhibitor.
Customers Who Bought This Also Bought