Z-Ile-Glu(OMe)-Thr-DL-Asp(OMe)-Fluoromethylketone
Supplier: Bachem Americas
Certificates
Certificates
About this item
Cell permeable inhibitor of caspase-8 and granzyme B. Z-IETD-FMK blocked CD95-induced apoptosis.
Customers Who Bought This Also Bought
Hydrogen peroxide 30% stabilized ACS, VWR Chemicals BDH®
Supplier: VWR International
$214.30 to $2,349.04
Combitips® advanced Eppendorf Quality Positive Displacement Pipette Tips, Eppendorf
Supplier: Eppendorf
$224.54
VWR® Absorbent Underpads with Waterproof Moisture Barrier
Supplier: VWR International
$240.47 to $8,653.27
VWR® Laboratory Bench and Table Protectors with Leakproof Moisture Barrier
Supplier: VWR International
$314.91
Sodium hydroxide 0.1 N in aqueous solution, VWR Chemicals BDH®
Supplier: VWR International
$78.90 to $874.91