Ac-VDVAD-CHO
Catalog #: N-1790.0005BA
Supplier: Bachem Americas
CAS Number: 194022-51-0
Specifications
- Size:5 mg
- Cat. no.:N-1790.0005BA
- Supplier No.:N-1790.0005
Specifications
About this item
Synonyms: Ac-Val-Asp-Val-Ala-Asp-aldehyde (pseudo acid)
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 543.574 g/mol
:Ac-VDVAD-CHO
Formula:C₂₃H₃₇N₅O₁₀
MW:543.574 g/mol
Storage Temperature:Freezer
MDL Number:MFCD01862574
CAS Number:194022-51-0
Customers Who Bought This Also Bought