Z-Leu-Leu-Tyr-alpha-Ketoaldehyde
Catalog #: N-1715.0005BA
Supplier: Bachem Americas
CAS Number:
Specifications
- CAS No.:204649-66-1
- Size:5 mg
- Storage conditions:–20 ± 5 °C
- MDL No.:MFCD03788098
- Protein/peptide name:Z-Leu-Leu-Tyr-alpha-keto aldehyde
- Molecular weight:553.66
- Sequence:Z-Leu-Leu-Tyr-α-keto aldehyde
- Molecular formula:C₃₀H₃₉N₃O₇
- Cat. no.:N-1715.0005BA
- Supplier No.:N-1715.0005
Specifications
About this item
See also Leupeptin (N-1000) and MG-132 (Calpain Inhibitor IV, N-1635).
Z-LLY-α-keto aldehyde (peptidyl glyoxal), previously shown to be an excellent inhibitor of the serine and cysteine proteases, was found to be an excellent inhibitor of the chymotryptic-like activity of proteasome exhibiting a Ki value of approximately 3.0 nM. Such inhibitor might be valuable in the characterization of the role of proteasome in normal and aberrant pathways of protein catabolism in the cell.
Customers Who Bought This Also Bought