Ac-Val-Glu-Ile-Asp-Aldehyde (Pseudo Acid)
Catalog #: N-1640.0005BA
Supplier: Bachem Americas
CAS Number:
Specifications
- CAS No.:319494-39-8
- Size:5 mg
- Storage conditions:–20 ± 5 °C
- MDL No.:MFCD01318861
- Protein/peptide name:Ac-Val-Glu-Ile-Asp-aldehyde (pseudo acid)
- Molecular weight:500.55
- Sequence:Ac-Val-Glu-Ile-Asp-aldehyde (pseudo acid)
- Molecular formula:C₂₂H₃₆N₄O₉
- Cat. no.:N-1640.0005BA
- Supplier No.:N-1640.0005
Specifications
About this item
AC-VEID-CHO, a specific inhibitor of caspase-6 (Mch2).
Customers Who Bought This Also Bought