Calpain Inhibitor IV
Catalog #: N-1635.0005BA
Supplier: Bachem Americas
CAS Number:
Specifications
- CAS No.:133407-82-6
- Size:5 mg
- Storage conditions:–20 ± 5 °C
- MDL No.:MFCD00674886
- Protein/peptide name:Calpain Inhibitor IV
- Molecular weight:475.63
- Sequence:Z-Leu-Leu-Leu-aldehyde
- Molecular formula:C₂₆H₄₁N₃O₅
- Cat. no.:N-1635.0005BA
- Supplier No.:N-1635.0005
Specifications
About this item
For additional calpain inhibitors, please see also our L-trans-Epoxysuccinyl Peptides.
Z-LLL-CHO (MG132) was shown to initiate neurite outgrowth in PC12 cells with an optimal concentration of 20 nM and to be a very strong inhibitor of 20S proteasome and of calpain.
Customers Who Bought This Also Bought