Biotinyl-epsilon-aminocaproyl-D-Phe-Pro-Arg-chloromethylketone
Catalog #: N-1565.0005BA
Supplier: Bachem Americas
CAS Number:
Specifications
- CAS No.:131104-10-4
- Size:5 mg
- Storage conditions:–20 ± 5 °C
- MDL No.:MFCD00270897
- Protein/peptide name:Biotinyl-epsilon-aminocaproyl-D-Phe-Pro-Arg-chloromethylketone
- Molecular weight:790.43
- Sequence:Biotinyl-ε-aminocaproyl-D-Phe-Pro-Arg-chloromethylketone
- Molecular formula:C₃₇H₅₆ClN₉O₆S
- Cat. no.:N-1565.0005BA
- Supplier No.:N-1565.0005
Specifications
About this item
Peptide and amino acid chloromethyl ketones (CMK's) are irreversible inhibitors of cysteine proteases, but they also react with serine proteases. For our amino acid CMK's (e.g. TPCK, TLCK) see the corresponding product family in the 'Amino acid' section.
Biotinylated derivative of PPACK (N-1065), a potent irreversible inhibitor of thrombin, tissue plasminogen activator (tPA), factor VIIa, and factor Xa. The inhibitor covalently binds to the enzymes and thereby inactivates them. Bound biotin allows reaction with avidin under a variety of different conditions. A procedure has for example been developed to remove active enzyme from zymogen preparations.
Customers Who Bought This Also Bought