Decanoyl-Arg-Val-Arg-Lys-chloromethylketone
Catalog #: N-1435.0005BA
Supplier: Bachem Americas
CAS Number:
Specifications
- CAS No.:534615-50-4
- Size:5 mg
- Storage conditions:–20 ± 5 °C
- MDL No.:MFCD02259677
- Protein/peptide name:Decanoyl-Arg-Val-Arg-Lys-chloromethylketone
- Molecular weight:744.42
- Sequence:Decanoyl-Arg-Val-Arg-Lys-chloromethylketone
- Molecular formula:C₃₄H₆₆ClN₁₁O₅
- Cat. no.:N-1435.0005BA
- Supplier No.:N-1435.0005
Specifications
About this item
Peptide and amino acid chloromethyl ketones (CMK's) are irreversible inhibitors of cysteine proteases, but they also react with serine proteases. For our amino acid CMK's (e.g. TPCK, TLCK) see the corresponding product family in the 'Amino acid' section.
The inhibitor Dec-RVRK-CMK has been used to characterize the specificity of furin-like proteases.
Customers Who Bought This Also Bought