Z-Asp(OtBu)-bromomethyl ketone
Catalog #: N-1425.1000BA
Supplier: Bachem Americas
CAS Number: 153088-76-7
Specifications
- Size:1 g
- Cat. no.:N-1425.1000BA
- Supplier No.:N-1425.1000
Specifications
About this item
Synonyms: Z-Asp(OtBu)-bromomethylketone
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 400.269 g/mol
:Z-Asp(OtBu)-bromomethyl ketone
Formula:C₁₇H₂₂BrNO₅
MW:400.269 g/mol
Storage Temperature:Freezer
MDL Number:MFCD00672490
CAS Number:153088-76-7
Customers Who Bought This Also Bought