Z-Phe-Lys-2,4,6-trimethylbenzoyloxymethylketone
Catalog #: N-1400.0025BA
Supplier: Bachem Americas
CAS Number: 118253-05-7
Specifications
- Size:25 mg
- Cat. no.:N-1400.0025BA
- Supplier No.:N-1400.0025
Specifications
About this item
Synonyms: Z-Phe-Lys-2,4
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 701,74 g/mol
Formula:C₃₆H₄₂F₃N₃O₈
Storage Temperature:Freezer
MDL Number:MFCD03788099
CAS Number:118253-05-7
Customers Who Bought This Also Bought