Ac-Leu-Val-Lys-Aldehyde
Supplier: Bachem Americas
Certificates
About this item
Formula:C₁₉H₃₆N₄O₄
MDL Number:MFCD00237496
CAS Number:147600-40-6
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 384.521 g/mol
:Ac-Leu-Val-Lys-Aldehyde
Storage Temperature:Freezer
Customers Who Bought This Also Bought