Z-Leu-Tyr-Chloromethyl Ketone
Catalog #: N-1255.0025BA
Supplier: Bachem Americas
CAS Number:
Specifications
- CAS No.:56979-35-2
- Size:25 mg
- Storage conditions:–20 ± 5 °C
- MDL No.:MFCD00238476
- Protein/peptide name:Z-Leu-Tyr-chloromethylketone
- Molecular weight:460.96
- Sequence:Z-Leu-Tyr-chloromethylketone
- Molecular formula:C₂₄H₂₉ClN₂O₅
- Cat. no.:N-1255.0025BA
- Supplier No.:N-1255.0025
Specifications
About this item
Peptide and amino acid chloromethyl ketones (CMK's) are irreversible inhibitors of cysteine proteases, but they also react with serine proteases. For our amino acid CMK's (e.g. TPCK, TLCK) see the corresponding product family in the 'Amino acid' section.
Z-LY-CMK, E.coli ClpP inhibitor.
Customers Who Bought This Also Bought