H-D-Pro-Phe-Arg-chloromethylketone
Catalog #: N-1210.0025BA
Supplier: Bachem Americas
CAS Number: 88546-74-1
Specifications
- Size:25 mg
- Cat. no.:N-1210.0025BA
- Supplier No.:N-1210.0025
Specifications
About this item
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 450.968 g/mol
Formula:C₂₁H₃₁ClN₆O₃
Storage Temperature:Freezer
MDL Number:MFCD18633509
CAS Number:88546-74-1
Customers Who Bought This Also Bought