Abz-Ala-Phe-Ala-Phe-Asp-Val-Phe-3-nitro-Tyr-Asp-OH Trifluoroacetate
Catalog #: M-2475.0001BA
Supplier: Bachem Americas
CAS Number:
Specifications
- CAS No.:143147-74-4
- Size:1 mg
- Storage conditions:–20 ± 5 °C
- MDL No.:MFCD18782561
- Protein/peptide name:Abz-Ala-Phe-Ala-Phe-Asp-Val-Phe-3-nitro-Tyr-Asp-OH Trifluoroacetate
- Molecular weight:1258.31
- Sequence:Abz-Ala-Phe-Ala-Phe-Asp-Val-Phe-3-nitro-Tyr-Asp-OH
- Molecular formula:C₆₂H₇₁N₁₁O₁₈
- Cat. no.:M-2475.0001BA
- Supplier No.:M-2475.0001
Specifications
About this item
Abz-AFAFDVY(NO₂)D, a fluorogenic (FRET) substrate for endoproteinase AspN, Asp-specific proteases from Staphylococcus aureus, Bacillus licheniformis and Streptomyces griseus.
Customers Who Bought This Also Bought