Ac-Glu-Asp(EDANS)-Lys-Pro-Ile-Leu-Phe-Phe-Arg-Leu-Gly-Lys(DABCYL)-Glu-NH₂ Trifluoroacetate
Catalog #: M-2295.0001BA
Supplier: Bachem Americas
CAS Number:
Specifications
- CAS No.:400716-78-1
- Size:1 mg
- Storage conditions:–20 ± 5 °C
- MDL No.:MFCD18782546
- Protein/peptide name:Ac-Glu-Asp(EDANS)-Lys-Pro-Ile-Leu-Phe-Phe-Arg-Leu-Gly-Lys(DABCYL)-Glu-NH₂ Trifluoroacetate
- Molecular weight:2132.52
- Sequence:Ac-Glu-Asp(EDANS)-Lys-Pro-Ile-Leu-Phe-Phe-Arg-Leu-Gly-Lys(DABCYL)-Glu-NH₂
- Molecular formula:C₁₀₄H₁₄₆N₂₄O₂₃S
- Cat. no.:M-2295.0001BA
- Supplier No.:M-2295.0001
Specifications
About this item
Ac-ED(edans)KPILFFRLGK(dabcyl)E-amide, a sensitive fluorescent (FRET) peptide substrate for cathepsin D. This enzyme can degrade extracellular matrix components and may facilitate the spread of tumor cells. High levels of active cathepsin D were found within senile plaques in brains of Alzheimer's patients.
Customers Who Bought This Also Bought