Ureidosuccinic acid
Supplier: Bachem Americas
Certificates
About this item
Synonyms: Carbamoyl-Asp-OH·magnesium salt/Carbamoyl-Asp-OH·dipotassium salt (1:1)
Formula:C₅H₈N₂O₅
MW:176.128 g/mol
MDL Number:MFCD03613594
CAS Number:13184-27-5
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 176.128 g/mol
:Ureidosuccinic acid
Storage Temperature:Refrigerator
EINECS:236-134-3
Customers Who Bought This Also Bought