Mca-Tyr-Val-Ala-Asp-Ala-Pro-Lys(Dnp)-OH Trifluoroacetate
Catalog #: M-2195.0001BA
Supplier: Bachem Americas
CAS Number:
Specifications
- CAS No.:189696-01-3
- Size:1 mg
- Storage conditions:–20 ± 5 °C
- MDL No.:MFCD18782539
- Protein/peptide name:Mca-Tyr-Val-Ala-Asp-Ala-Pro-Lys(Dnp)-OH Trifluoroacetate
- Molecular weight:1145.15
- Sequence:Mca-Tyr-Val-Ala-Asp-Ala-Pro-Lys(Dnp)-OH
- Molecular formula:C₅₃H₆₄N₁₀O₁₉
- Cat. no.:M-2195.0001BA
- Supplier No.:M-2195.0001
Specifications
About this item
Specific highly fluorescent FRET substrate for the determination of caspase-1 (ICE) and caspase-1-like enzyme activities. Cleavage of Mca-YVADAPK(Dnp) at the P1 Asp residue results in a continuous fluorescent assay monitored at an emission wavelength of 392 nm. Caspase-3 (apopain, CPP-32) is only weakly active on this substrate. Both caspase-1 and caspase-3 are involved in apoptosis. A specific substrate for renal ACE 2.
Customers Who Bought This Also Bought