Nɑ-Tosyl-L-arginine methyl ester hydrochloride
Catalog #: M-1745.0025BA
Supplier: Bachem Americas
CAS Number: 1784-03-8
Specifications
- Size:25 g
- Cat. no.:M-1745.0025BA
- Supplier No.:M-1745.0025
Specifications
About this item
Synonyms: TAME,Nɑ-p-Toluenesulfonyl-L-arginine Methyl Ester Hydrochloride,Nɑ-p-Toluenesulphonyl-L-arginine Methyl Ester Hydrochloride,Tos-Arg-OMe.HCl
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 378.87 g/mol
Formula:C₁₄H₂₂N₄O₄S·HCl
MW:378.87 g/mol
Storage Temperature:Freezer
MDL Number:MFCD00012578
CAS Number:1784-03-8
EINECS:217-235-1
Customers Who Bought This Also Bought