Ac-Lys(Ac)-D-Ala-D-lactic acid
Catalog #: M-1330.0050BA
Supplier: Bachem Americas
CAS Number:
Specifications
- CAS No.:65882-12-4
- Size:50 mg
- Storage conditions:–20 ± 5 °C
- MDL No.:MFCD00038705
- Protein/peptide name:Ac-Lys(Ac)-D-Ala-D-lactic acid
- Molecular weight:373.41
- Sequence:Ac-Lys(Ac)-D-Ala-D-lactic acid
- Molecular formula:C₁₆H₂₇N₃O₇
- Cat. no.:M-1330.0050BA
- Supplier No.:M-1330.0050
Specifications
About this item
See also F-3260.
Ac₂Lys-D-Ala-D-Lac-OH is a depsipeptide substrate for penicillin-sensitive D-alanine carboxypeptidases (DD-carboxypeptidases).
Customers Who Bought This Also Bought