(S)-Methyl-6-amino-2-(((benzyloxy)carbonyl)amino)hexanoate hydrochloride
Supplier: Bachem Americas
Certificates
About this item
Synonyms: (S)-Methyl 6-amino-2-(((benzyloxy)carbonyl)amino)hexanoate hydrochloride
Formula:C₁₅H₂₃ClN₂O₄
MW:330.809 g/mol
MDL Number:MFCD00077025
CAS Number:26348-68-5
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 330.809 g/mol
:(S)-Methyl-6-amino-2-(((benzyloxy)carbonyl)amino)hexanoate hydrochloride
Storage Temperature:Freezer
Customers Who Bought This Also Bought