N feminine(1)U(1/2)oOAeAO-
Catalog #: M-1115.0005BA
Supplier: Bachem Americas
CAS Number: 20845-02-7
Specifications
- Size:5 mg
- Cat. no.:M-1115.0005BA
- Supplier No.:M-1115.0005
Specifications
About this item
Synonyms: H-Asp-Arg-Val-Tyr-Ile-His-Pro-Phe-His-Leu-Leu-Val-Tyr-Ser-OH
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 1759.046 g/mol
:N feminine(1)U(1/2)oOAeAO-
Formula:C₈₅H₁₂₃N₂₁O₂₀
MW:1759.046 g/mol
Storage Temperature:Freezer
MDL Number:MFCD00133798
CAS Number:20845-02-7
Customers Who Bought This Also Bought