Suc-Tyr-Val-Ala-Asp-Pna
Catalog #: L-1830.0025BA
Supplier: Bachem Americas
CAS Number: 208264-84-0
Specifications
- Size:25 mg
- Cat. no.:L-1830.0025BA
- Supplier No.:L-1830.0025
Specifications
About this item
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 686.675 g/mol
:Suc-Tyr-Val-Ala-Asp-Pna
Formula:C₃₁H₃₈N₆O₁₂
Storage Temperature:Freezer
MDL Number:MFCD01318836
CAS Number:208264-84-0
Customers Who Bought This Also Bought