H-Asp(pNA)-OH·HCl
Catalog #: L-1690.0250BA
Supplier: Bachem Americas
CAS Number: 201732-79-8
Specifications
- Size:250 mg
- Cat. no.:L-1690.0250BA
- Supplier No.:L-1690.0250
Specifications
About this item
Synonyms: H-Asp(pNA)-OH·HCl
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 289.672 g/mol
:H-Asp(pNA)-OH·HCl
Formula:C₁₀H₁₂ClN₃O₅
MW:289.672 g/mol
Storage Temperature:Freezer
MDL Number:MFCD00236807
CAS Number:201732-79-8
Customers Who Bought This Also Bought