(S)-3-Amino-4-((4-nitrophenyl)amino)-4-oxobutanoic acid hydrochloride
Catalog #: L-1525.0001BA
Supplier: Bachem Americas
CAS Number: 154564-03-1
Specifications
- Size:1 g
- Cat. no.:L-1525.0001BA
- Supplier No.:L-1525.0001
Specifications
About this item
Synonyms: H-Asp-pNA·HCl
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 289.672 g/mol
:(S)-3-Amino-4-((4-nitrophenyl)amino)-4-oxobutanoic acid hydrochloride
Formula:C₁₀H₁₂ClN₃O₅
MW:289.672 g/mol
Storage Temperature:Freezer
MDL Number:MFCD00237823
CAS Number:154564-03-1
Customers Who Bought This Also Bought