Boc-Ala-Ala-Asp-pNA
Catalog #: L-1165.0050BA
Supplier: Bachem Americas
CAS Number:
Specifications
- CAS No.:201732-83-4
- Size:50 mg
- Storage conditions:–20 ± 5 °C
- MDL No.:MFCD00237526
- Protein/peptide name:Boc-Ala-Ala-Asp-pNA
- Molecular weight:495.49
- Sequence:Boc-Ala-Ala-Asp-pNA
- Molecular formula:C₂₁H₂₉N₅O₉
- Cat. no.:L-1165.0050BA
- Supplier No.:L-1165.0050
Specifications
About this item
Boc-AAD-pNA, a chromogenic substrate for human and murine granzyme B.
Customers Who Bought This Also Bought