L-Isoleucine-β-naphthylamide
Catalog #: K-1375.0001BA
Supplier: Bachem Americas
CAS Number: 732-84-3
Specifications
- Size:1 g
- Cat. no.:K-1375.0001BA
- Supplier No.:K-1375.0001
Specifications
About this item
Synonyms: H-Ile-βNA
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 256.349 g/mol
Formula:C₁₆H₂₀N₂O
MW:256.349 g/mol
Storage Temperature:Ambient
MDL Number:MFCD00055912
CAS Number:732-84-3
Customers Who Bought This Also Bought