L-Histidine-β-naphthylamide
Catalog #: K-1350.0250BA
Supplier: Bachem Americas
CAS Number: 7424-15-9
Specifications
- Size:250 mg
- Cat. no.:K-1350.0250BA
- Supplier No.:K-1350.0250
Specifications
About this item
Synonyms: H-His-betaNA
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 280.331 g/mol
:L-Histidine-β-naphthylamide
Formula:C₁₆H₁₆N₄O
MW:280.331 g/mol
Storage Temperature:Ambient
MDL Number:MFCD00051166
CAS Number:7424-15-9
EINECS:231-052-4
Customers Who Bought This Also Bought