Z-Arg-Arg-betaNA Acetate
50 mg
Catalog # K-1170.0050BA
Jump to:
Discontinued
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Z-RR-βNA, specific substrate of maximal sensitivity for cathepsin B, free from interference by cathepsin H.
Specifications
Specifications
- Size:50 mg
- Cat. No.:K-1170.0050BA
- Molecular formula:C₃₀H₃₉N₉O₄
- Protein/peptide name:Z-Arg-Arg-betaNA Acetate
- Storage conditions:–20 ± 5 °C
- MDL No.:MFCD03788092
- Sequence:Z-Arg-Arg-βNA
- Molecular weight:589.70
- CAS No.:76474-52-7
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



