H-Ala-βNA
Supplier: Bachem Americas
Certificates
About this item
Formula:C₁₃H₁₄N₂O
MDL Number:MFCD00064969
CAS Number:720-82-1
:H-Ala-βNA
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 214.27 g/mol
Storage Temperature:Ambient
EINECS:211-956-5
Customers Who Bought This Also Bought