Bz-Arg-Gly-Phe-Phe-Pro-4MbetaNA · HCl
Catalog #: J-1065.0050BA
Supplier: Bachem Americas
CAS Number:
Specifications
- CAS No.:201928-98-5
- Size:50 mg
- Storage conditions:–20 ± 5 °C
- MDL No.:MFCD00155570
- Protein/peptide name:Bz-Arg-Gly-Phe-Phe-Pro-4MbetaNA · HCl
- Molecular weight:918.49
- Sequence:Bz-Arg-Gly-Phe-Phe-Pro-4MβNA · HCl
- Molecular formula:C₄₉H₅₅N₉O₇ · HCl
- Cat. no.:J-1065.0050BA
- Supplier No.:J-1065.0050
Specifications
About this item
Bz-RGFFP-MNA is used for the colorimetric or fluorometric determination of cathepsin D. Excitation at ca. 335 nm, emission at ca. 410 nm.
Customers Who Bought This Also Bought