Ac-Arg-Gly-Lys-AMC Trifluoroacetate
Catalog #: I-1990.0005BA
Supplier: Bachem Americas
CAS Number:
Specifications
- CAS No.:660846-99-1
- Size:5 mg
- Storage conditions:–20 ± 5 °C
- MDL No.:MFCD18782522
- Protein/peptide name:Ac-Arg-Gly-Lys-AMC Trifluoroacetate
- Molecular weight:558.64
- Sequence:Ac-Arg-Gly-Lys-AMC
- Molecular formula:C₂₆H₃₈N₈O₆
- Cat. no.:I-1990.0005BA
- Supplier No.:I-1990.0005
Specifications
About this item
See also Boc-Lys(Ac)-AMC (I-1875) and Boc-Lys(Tfa)-AMC (I-1985).
Control for the two step histone deacetylase assay with Ac-Arg-Gly-Lys(Ac)-AMC (I-1925). Ac-RGK-AMC corresponds to the product of the deacetylase reaction, which is subsequently cleaved by trypsin.
Customers Who Bought This Also Bought