Ac-Gly-Ala-Lys-AMC Trifluoroacetate
Catalog #: I-1980.0005BA
Supplier: Bachem Americas
CAS Number:
Specifications
- Size:5 mg
- Storage conditions:–20 ± 5 °C
- MDL No.:MFCD18782521
- Protein/peptide name:Ac-Gly-Ala-Lys-AMC Trifluoroacetate
- Molecular weight:473.53
- Sequence:Ac-Gly-Ala-Lys-AMC
- Molecular formula:C₂₃H₃₁N₅O₆
- Cat. no.:I-1980.0005BA
- Supplier No.:I-1980.0005
Specifications
About this item
See also Boc-Lys(Ac)-AMC (I-1875) and Boc-Lys(Tfa)-AMC (I-1985).
Ac-GAK-AMC, control peptide for protease-coupled histone deacetylase (HDAC) assay with Ac-Gly-Ala-Lys(Ac)-AMC (I-1975).
Customers Who Bought This Also Bought