Boc-Lys(Ac)-AMC
Catalog #: I-1875.0250BA
Supplier: Bachem Americas
CAS Number: 233691-67-3
Specifications
- Size:250 mg
- Cat. no.:I-1875.0250BA
- Supplier No.:I-1875.0250
Specifications
About this item
Synonyms: (S)-tert-Butyl (6-acetamido-1-((4-methyl-2-oxo-2H-chromen-7-yl)amino)-1-oxohexan-2-yl)carbamate
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 445.516 g/mol
:Boc-Lys(Ac)-AMC
Formula:C₂₃H₃₁N₃O₆
MW:445.516 g/mol
Storage Temperature:Freezer
MDL Number:MFCD02683946
CAS Number:233691-67-3
Customers Who Bought This Also Bought