H-Asp-AMC
Supplier: Bachem Americas
Certificates
About this item
Formula:C₁₄H₁₄N₂O₅
MDL Number:MFCD00133439
CAS Number:219138-13-3
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 290.275 g/mol
:H-Asp-AMC
Storage Temperature:Freezer
Customers Who Bought This Also Bought