Ac-Asp-Glu-Val-Asp-AFC
Catalog #: I-1725.0005BA
Supplier: Bachem Americas
CAS Number:
Specifications
- CAS No.:201608-14-2
- Size:5 mg
- Storage conditions:–20 ± 5 °C
- MDL No.:MFCD01310970
- Protein/peptide name:Ac-Asp-Glu-Val-Asp-AFC
- Molecular weight:729.62
- Sequence:Ac-Asp-Glu-Val-Asp-AFC
- Molecular formula:C₃₀H₃₄F₃N₅O₁₃
- Cat. no.:I-1725.0005BA
- Supplier No.:I-1725.0005
Specifications
About this item
Ac-DEVD-AFC is a sensitive fluorescent substrate for the assay of caspase-3 activity. The trifluoromethyl substituent of the fluorophore improves the membrane permeability of the substrate. CAS-Number (net) 141258-58-4 .
Customers Who Bought This Also Bought