1-[4-(2-Hydroxyethyl)piperazin-1-yl]propan-2-ol
Catalog #: I-1710.0005BA
Supplier: Bachem Americas
CAS Number: 219137-91-4
Specifications
- Size:5 mg
- Cat. no.:I-1710.0005BA
- Supplier No.:I-1710.0005
Specifications
About this item
Synonyms: Z-Val-Ala-Asp-AMC
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 188.271 g/mol
:1-[4-(2-Hydroxyethyl)piperazin-1-yl]propan-2-ol
Formula:C₉H₂₀N₂O₂
MW:188.271 g/mol
Storage Temperature:Freezer
MDL Number:MFCD01318828
CAS Number:219137-91-4
Customers Who Bought This Also Bought