Ac-Tyr-Val-Ala-Asp-AFC
Catalog #: I-1705.0005BA
Supplier: Bachem Americas
CAS Number: 219137-85-6
Specifications
- Size:5 mg
- Cat. no.:I-1705.0005BA
- Supplier No.:I-1705.0005
Specifications
About this item
Synonyms: Ac-Tyr-Val-Ala-Asp-AFC
:Ac-Tyr-Val-Ala-Asp-AFC
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 481.453 g/mol
Formula:C₂₂H₂₅F₂N₃O₇
Storage Temperature:Freezer
MDL Number:MFCD01318881
CAS Number:219137-85-6
Customers Who Bought This Also Bought