Boc-Gln-Arg-Arg-Mca
Catalog #: I-1655.0025BA
Supplier: Bachem Americas
CAS Number: 109376-05-8
Specifications
- Size:25 mg
- Cat. no.:I-1655.0025BA
- Supplier No.:I-1655.0025
Specifications
About this item
Synonyms: Boc-Gln-Arg-Arg-AMC
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 775.865 g/mol
:Boc-Gln-Arg-Arg-Mca
Formula:C₃₄H₅₃N₁₁O₁₀
MW:775.865 g/mol
Storage Temperature:Freezer
MDL Number:MFCD03788110
CAS Number:109376-05-8
Customers Who Bought This Also Bought